Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPZ1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568871
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568871 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568871 Supplier Invitrogen™ Supplier No. PA568871
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Mouse: 100%.

Spermatogenic leucine zipper protein 1 is a member of the basic helix-loop-helix (bHLH) family of transcription factors which is expressed specifically in the testis and epididymis. Studies show that SPZ1 acts downstream of mitogen-activated protein kinase (MAPK), an extracellular signal-regulated kinase 1/2 to up-regulate cell proliferation and tumorigenesis. Also, through an interaction with proliferating cell nuclear antigen (PCNA) promoter, SPZ1 induced cell proliferation concomitant with an increase in PCNA gene expression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPZ1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Spz1
Gene Accession No. Q99MY0
Gene Alias 1700027M20Rik; AI429130; bhlh-zip transcription factor SPZ1; NYD-TSP1; PPP1R148; protein phosphatase 1, regulatory subunit 148; spermatogenic leucine zipper 1; spermatogenic leucine zipper protein 1; Spermatogenic Zip 1; SPZ1; testis secretory sperm-binding protein Li 232m; testis-specific protein 1; Testis-specific protein NYD-TSP1; TSP1
Gene Symbols Spz1
Host Species Rabbit
Immunogen synthetic peptide directed towards the C terminal region of mouse SPZ1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79401
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.