Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SQLE Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553712
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553712 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553712 Supplier Invitrogen™ Supplier No. PA553712
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DGRKVTVIER DLKEPDRIVG EFLQPGGYHV LKDLGLGDTV EGLDAQVVNG YMIHDQESKS EVQIPYPLSE NNQVQSGRAF HHGRFIMSLR KAAMAEPNAK FIEGVVLQLL EEDDVVMG Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 81%.

SQLE, also named as ERG1, SE and SM, belongs to the squalene monooxygenase family. It catalyzes the first oxygenation step in cholesterol synthesis, acting on squalene before cyclization into the basic steroid structure. SQLE may serve as a flux-controlling enzyme beyond 3-hydroxy-3-methylglutaryl-coenzyme A reductase (HMGR, considered as rate limiting). It is also post translationally regulated by cholesterol-dependent proteasomal degradation. The MW of SQLE is about 64 kDa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SQLE
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SQLE
Gene Accession No. Q14534
Gene Alias AI323792; Erg1; FLJ30795; SE; Sqle; Squalene epoxidase; squalene monooxygenase; Squalene monooxygenase-like protein
Gene Symbols SQLE
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SQLE. Recombinant protein control fragment (Product #RP-91198).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6713
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.