Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SQLE Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554265
Description
Immunogen sequence: ELHAPLTVVA DGLFSKFRKS LVSNKVSVSS HFVGFLMKNA PQFKANHAEL ILANPSPVLI YQISSSETRV LVDIRGEMPR NLREYMVEKI YPQIPDHLKE PFLEATDNSH LRSMPASFLP PSSVKKRGVL LLGDA Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 86%.
SQLE, also named as ERG1, SE and SM, belongs to the squalene monooxygenase family. It catalyzes the first oxygenation step in cholesterol synthesis, acting on squalene before cyclization into the basic steroid structure. SQLE may serve as a flux-controlling enzyme beyond 3-hydroxy-3-methylglutaryl-coenzyme A reductase (HMGR, considered as rate limiting). It is also post translationally regulated by cholesterol-dependent proteasomal degradation. The MW of SQLE is about 64 kDa.
Specifications
SQLE | |
Polyclonal | |
Unconjugated | |
SQLE | |
AI323792; Erg1; FLJ30795; SE; Sqle; Squalene epoxidase; squalene monooxygenase; Squalene monooxygenase-like protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6713 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q14534 | |
SQLE | |
Recombinant protein corresponding to Human SQLE. Recombinant protein control fragment (Product #RP-91199). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction