Learn More
MilliporeSigma SQPAK™ Care A Cleaner
Catalog No. SQPAKACARE Shop All MilliporeSigma Products
Change view
Click to view available options
:
Type: Cleaner
Catalog No.
SQPAKACARE Type: Cleaner
1 options
Catalog No. SQPAKACARE Supplier MilliporeSigma Supplier No. SQPAKACARE
Only null left
Add to Cart
Add to Cart
Anti-scaling agent for use with Milli-Q™ SQ 2Series RO production module
  • Preventive anti-scaling agent to clean hydraulic parts of pure RO production station.
  • For use on the pure RO production station of Milli-Q™ SQ 2Series configurations, including Milli-Q™ SR 240/240L/240XL and Milli-Q™ SQ 240C/240CV systems.
  • Easy to use: simply add cleaning agent to Milli-Q™ SQ Cleaning tool ZSQ240TT0 (not included).
TRUSTED_SUSTAINABILITY
Specifications
Type Cleaner