Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SQRDL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553549
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553549 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553549 Supplier Invitrogen™ Supplier No. PA553549
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TSPVADAAGW VDVDKETLQH RRYPNVFGIG DCTNLPTSKT AAAVAAQSGI LDRTISVIMK NQTPTKKYDG YTSCPLVTGY NRVILAEFDY KAEPLETFPF DQSKERLSMY LMKADLMPFL YWNMMLRGYW G Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 93%.

SQRDL (Sulfide: quinone oxidoreductase, mitochondrial) catalyzes the oxidation of hydrogen sulfide, with the help of a quinone. Although the SQRDL precursor protein displays a molecular weight of 50 kDa, a prominent band at about 46 kDa is detected by western blot. This discrepancy reflects the cleavage of an N-terminal targeting sequence of 4 kDa during the import of the protein into the mitochondria. SQRDL is one of the enzymes involved in H2S signaling in a discrete population of neurons, oligodendrocytes, and endothelial cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SQRDL
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.30 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Sqor
Gene Accession No. Q9R112, Q9Y6N5
Gene Alias 0610039J17Rik; 4930557M22Rik; Cc2-17; CGI-44; flavo-binding protein; PRO1975; SQOR; Sqrdl; sulfide dehydrogenase like; Sulfide dehydrogenase-like; Sulfide quinone oxidoreductase; sulfide quinone reductase-like (yeast); sulfide:quinone oxidoreductase, mitochondrial
Gene Symbols Sqor
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SQRDL. Recombinant protein control fragment (Product #RP-89842).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58472, 59010
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.