Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SQRDL Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559637
Description
Immunogen sequence: LIIALGIQLD YEKIKGLPEG FAHPKIGSNY SVKTVEKTWK ALQDFKEGNA IFTFPNTPVK CAGAPQKIMY LSEAYFRKTG KRSKANIIFN TSLGAIFGVK KYADALQEII QERNLTVNYK KNLIEVRADK QEAVFENLD Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 94%.
SQRDL (Sulfide: quinone oxidoreductase, mitochondrial) catalyzes the oxidation of hydrogen sulfide, with the help of a quinone. Although the SQRDL precursor protein displays a molecular weight of 50 kDa, a prominent band at about 46 kDa is detected by western blot. This discrepancy reflects the cleavage of an N-terminal targeting sequence of 4 kDa during the import of the protein into the mitochondria. SQRDL is one of the enzymes involved in H2S signaling in a discrete population of neurons, oligodendrocytes, and endothelial cells.
Specifications
SQRDL | |
Polyclonal | |
Unconjugated | |
Sqor | |
0610039J17Rik; 4930557M22Rik; Cc2-17; CGI-44; flavo-binding protein; PRO1975; SQOR; Sqrdl; sulfide dehydrogenase like; Sulfide dehydrogenase-like; Sulfide quinone oxidoreductase; sulfide quinone reductase-like (yeast); sulfide:quinone oxidoreductase, mitochondrial | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
58472 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y6N5 | |
Sqor | |
Recombinant protein corresponding to Human SQRDL. Recombinant protein control fragment (Product #RP-89841). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction