Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SQRDL Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5112723
Description
SQRDL Polyclonal Antibody for Western Blot, IHC (P), ELISA
SQRDL (Sulfide: quinone oxidoreductase, mitochondrial) catalyzes the oxidation of hydrogen sulfide, with the help of a quinone. Although the SQRDL precursor protein displays a molecular weight of 50 kDa, a prominent band at about 46 kDa is detected by western blot. This discrepancy reflects the cleavage of an N-terminal targeting sequence of 4 kDa during the import of the protein into the mitochondria. SQRDL is one of the enzymes involved in H2S signaling in a discrete population of neurons, oligodendrocytes, and endothelial cells.
Specifications
SQRDL | |
Polyclonal | |
Unconjugated | |
Sqor | |
0610039J17Rik; 4930557M22Rik; Cc2-17; CGI-44; flavo-binding protein; PRO1975; SQOR; Sqrdl; sulfide dehydrogenase like; Sulfide dehydrogenase-like; Sulfide quinone oxidoreductase; sulfide quinone reductase-like (yeast); sulfide:quinone oxidoreductase, mitochondrial | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
58472 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
1.5 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9Y6N5 | |
Sqor | |
Recombinant Human Sulfide:quinone oxidoreductase, mitochondrial protein (211-450AA). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction