Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SQSTM1 Monoclonal Antibody (OTI4G9), TrueMAB™, OriGene
SDP

Catalog No. 502541773
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-1773 30 μL
50-167-3212 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-1773 Supplier OriGene Supplier No. TA502132S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SQSTM1 Monoclonal antibody specifically detects SQSTM1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen SQSTM1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI4G9
Concentration 0.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Sqstm1
Gene Accession No. Q13501
Gene Alias A170; EBI3-associated protein of 60 kDa; EBI3-associated protein p60; EBIAP; FTDALS3; hypothetical protein 1; ORCA; OSF-6; Osi; OSIL; oxidative stress induced; oxidative stress induced like; p60; p62; p62B; PDB3; phosphotyrosine independent ligand for the Lck SH2 domain p62; phosphotyrosine-independent ligand for the Lck SH2 domain of 62 kDa; PKC-zeta-interacting protein; protein kinase C-zeta-interacting protein; sb:cb621; Sequestosome 1; sequestosome-1; Sqstm1; STAP; STONE14; ubiquitin-binding protein p62; Unknown (protein for MGC:127197); zgc:85784; ZIP; ZIP3
Host Species Mouse
Immunogen Full length human recombinant protein of human SQSTM1 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8878
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.