Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SQSTM1 Monoclonal Antibody (OTI6A7), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA502239S


Description
SQSTM1 Monoclonal antibody specifically detects SQSTM1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
SQSTM1 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q13501 | |
Mouse | |
Affinity chromatography | |
RUO | |
8878 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG2a |
Flow Cytometry, Immunocytochemistry, Western Blot | |
OTI6A7 | |
Unconjugated | |
Sqstm1 | |
A170; EBI3-associated protein of 60 kDa; EBI3-associated protein p60; EBIAP; FTDALS3; hypothetical protein 1; ORCA; OSF-6; Osi; OSIL; oxidative stress induced; oxidative stress induced like; p60; p62; p62B; PDB3; phosphotyrosine independent ligand for the Lck SH2 domain p62; phosphotyrosine-independent ligand for the Lck SH2 domain of 62 kDa; PKC-zeta-interacting protein; protein kinase C-zeta-interacting protein; sb:cb621; Sequestosome 1; sequestosome-1; Sqstm1; STAP; STONE14; ubiquitin-binding protein p62; Unknown (protein for MGC:127197); zgc:85784; ZIP; ZIP3 | |
Full length human recombinant protein of human SQSTM1 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction