Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SQSTM1 Recombinant Rabbit Monoclonal Antibody (PS00-61)
GREENER_CHOICE

Catalog No. FEMA550706
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550706 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550706 Supplier Invitrogen™ Supplier No. MA550706
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive control: PC-12 cell lysate, RAW264.7 cell lysate, PANC-1 cell lysate, mouse liver tissue lysate, rat brain tissue lysate, SH-SY5Y cell lysate, HeLa cell lysate, Hela-si-NT cell lysate, Hela-si-p62#1 cell lysate, Hela-si-p62#2 cell lysate, human colon carcinoma tissue, A549. Predicted band size: 48 kDa Subcellular Location: Cytoplasm, Endoplasmic reticulum, Endosome, Lysosome, Nucleus.

This gene encodes a multifunctional protein that binds ubiquitin and regulates activation of the nuclear factor kappa-B signaling pathway. The protein functions as a scaffolding/adaptor protein in concert with TNF receptor-associated factor 6 to mediate activation of NF-kB in response to upstream signals. Alternatively spliced transcript variants encoding either the same or different isoforms have been identified for this gene. Mutations in this gene result in sporadic and familial Paget disease of bone.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SQSTM1
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone PS00-61
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Sqstm1
Gene Accession No. O08623, Q13501, Q64337
Gene Alias A170; EBI3-associated protein of 60 kDa; EBI3-associated protein p60; EBIAP; FTDALS3; hypothetical protein 1; ORCA; OSF-6; Osi; OSIL; oxidative stress induced; oxidative stress induced like; p60; p62; p62B; PDB3; phosphotyrosine independent ligand for the Lck SH2 domain p62; phosphotyrosine-independent ligand for the Lck SH2 domain of 62 kDa; PKC-zeta-interacting protein; protein kinase C-zeta-interacting protein; sb:cb621; Sequestosome 1; sequestosome-1; Sqstm1; STAP; STONE14; ubiquitin-binding protein p62; Unknown (protein for MGC:127197); zgc:85784; ZIP; ZIP3
Gene Symbols Sqstm1
Host Species Rabbit
Immunogen Synthetic peptide within Human SQSTM1/ p62 aa 400 to the C-terminus.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 113894, 18412, 8878
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.