Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Squalane, 99%, Thermo Scientific Chemicals

Catalog No. AC215355000
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-01-3
InChI Key PRAKJMSDJKAYCZ-UHFFFAOYSA-N
Synonym squalane, perhydrosqualene, dodecahydrosqualene, cosbiol, vitabiosol, spinacane, robane, hexamethyltetracosane, tetracosane, 2,6,10,15,19,23-hexamethyl, hexamethyl tetracosane
PubChem CID 8089
IUPAC Name 2,6,10,15,19,23-hexamethyltetracosane
SMILES CC(C)CCCC(C)CCCC(C)CCCCC(C)CCCC(C)CCCC(C)C

Specifications

Acidity 0.2mg KOH/g max.
Color Colorless
Density 0.8085g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C30H62
Refractive Index 1.4510 to 1.4525 (20°C, 589nm)
Quantity 500 mL
Saponification Value 1mg KOH/g max.
Molecular Weight (g/mol) 422.826
Formula Weight 422.82
Percent Purity ≥98.5% (GC)
Physical Form Liquid
Chemical Name or Material Squalane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.