Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Squalane, NF, 97-102%, Spectrum™ Chemical

Click to view available options
Quantity:
100 mL
500 mL
12 kg
20 L
Packaging:
Amber Glass Bottle
Description
Spectrum™ Chemical Squalane, NF is used as an emollient and moisturizer. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
CAS Max % | 1 |
Boiling Point | 212°C |
Ignition Residue | 0.005 |
Packaging | Amber Glass Bottle |
Refractive Index | 1.4510-1.4525 |
Quantity | 500 mL |
Saponification Value | 2 |
Specific Gravity | 0.807-0.810 |
Percent Purity | 97 to 102% |
Grade | NF |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction