Learn More
Description
- Square reagent bottles - Wide mouth square HDPE reagent bottles with polypropylene caps, designed for easy handling and efficient storage in laboratory environments.
- Leak-proof tested - Rigorously tested to ensure secure sealing under pressure, preventing spills during storage or transport.
- Chemical resistance for versatile use - Made from HDPE, these bottles are ideal for storing a wide range of chemicals and reagents.
- Laboratory and industrial applications - Perfect for chemical storage, liquid handling, and mixing in laboratories, research, and industrial settings.
- Translucent and durable design - Lightweight, translucent construction provides visibility of contents, while the square shape optimizes storage space.
- Designed for chemical storage and transport, these leak-proof, chemical-resistant bottles are ideal for laboratory and industrial applications. Non-sterile, -100°C to 120°C.
Specifications
Specifications
| Capacity (Metric) | 250 mL |
| Capacity (English) | 8.45 oz. |
| Color | Translucent |
| Material | HDPE |
| Sterility | Non-sterile |
| Closure Included | Yes |
| Graduated | No |
| Mouth | Wide |
| Product Type | Reagent Bottle |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.