Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ Squids™ 3158 Coil Tool Lanyard with Clamp

Catalog No. p-7163156 Shop All Ergodyne Products

Squids™ 3158 Coil Tool Lanyard with Clamp is compact and used to tether small hand tools, which prevents falling objects when working at heights.

  • Low-profile durable plastic coil to withstand even the toughest conditions
  • Easily attaches to hard hats and other small hand tools for safekeeping
  • Coiled lanyard design prevents snag and tangle hazards
  • Quick connect clamp attaches securely to fabric
  • Tethering loop to securely connect to an anchor or tool attachment
  • Certified to a 2:1 safety factor

Specifications

Length (Metric) 30 to 122 cm
Load Bearing Capacity (Metric) 0.9 kg
Load Bearing Capacity (English) 2 lb.
Length (English) 12 to 48 in.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.