Learn More
Ergodyne™ Squids™ 3410 Grabber Anchor with Suction Cup Mount
Catalog No. 19820315 Shop All Ergodyne Products
Change view
Click to view available options
Color: Black
Catalog No.
19-820-315 Color: Black
1 options
Catalog No. 19-820-315 Supplier Ergodyne™ Supplier No. 19321
Squids™ 3410 Grabber Anchor with Suction Cup Mount creates a modular system to move grabbers from belts, to lockers, and more.
  • Detachable, usable with carabiners, D-rings, etc.
  • Breakaway feature
  • Dielectric, non-conductive material
  • Suction cup base creates vacuum on smooth flat surfaces
Specifications
Includes Suction Cup Mount