Description
- Detachable, usable with carabiners, D-rings, etc.
- Breakaway feature
- Dielectric, non-conductive material
- Magnetic base attracts to ferromagnetic surfaces
- Encapsulated design houses magnet
- Smooth back plate provides a barrier against mounting surface
- Rubberized cap provides a friction ring around base to prevent it from sliding
Specifications
Specifications
| Color | Black |
| Includes | Magnet Mount |