Learn More
Ergodyne™ Squids™ 3760 Water Resistant Phone Pouch and Trap
Squids™ Water Resistant Phone Pouch and Trap makes it easy to carry and use your cell phone on the job, while preventing dropped objects when working at heights.
Supplier:  Ergodyne™ 19762
 View more versions of this product
Catalog No. 19-820-346
Only null left
Add to Cart
Description
Description
  • Approved to the ANSI/ISEA 121-2018 standard
  • Water resistant main material protects cell phone from rain, snow, dirt, dust, and sand
  • Clear touchscreen material allows for full screen and camera functions
  • Heavy-duty zipper securely holds cell phones inside the pouch
  • Steel grommet/ring can be used to easily connect a tool lanyard
  • Tarpaulin reinforced side material for durability
Specifications
Specifications
Standard
ANSI/ISEA 121-2018 standard
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.