Learn More
Ergodyne™ Squids™ 3796 Power Tool Bracket with Drill and Impact Driver Tool Trap™
Shop All Ergodyne ProductsDescription
- Stainless-steel bracket with built-in connection point for tool tethering is corrosion-resistant to withstand weathering
- Tool bracket design discretely fastens to open screw port on cordless drills/drivers
- Ideal for smaller, cylindrical drills, impact drivers, or other power tools
- Ambidextrous design can be flipped to either side of the tool attaching to open screw ports meant for accessory belt clips
- Fasteners include alloy steel M2.5 (8 mm length) socket cap head, 2 stainless steel M3 (8 mm and 12 mm lengths) Philips pan heads, and 3 stainless steel M4 (8 mm, 10 mm, 12 mm lengths) Philips pan heads
- Maximum tethering capacity of 2.5 lb. (1.13 kg) for the M2.5 fastener and 3 lb. (1.36 kg) for M3 and M4 fasteners
Specifications
Specifications
| Material | Stainless Steel |
| Color | Blue |
| Includes | 1 bracket connection point, 6 fasteners with locking washers, 2 product labels |
| Certifications/Compliance | ANSI/ISEA 121-2018 standard |