Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SR 0987
GREENER_CHOICE

Catalog No. 586810
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
58-681-0 10 mg
58-685-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 58-681-0 Supplier Tocris Bioscience™ Supplier No. 5868/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

T cell-specific RORγ (RORγt) agonist

SR 0987 is a T cell-specific RORγ (RORγt) agonist (EC50 = 800 nM in a gene reporter assay). Induces IL-17 expression and decreases PD-1 expression in T cells in vitro.

Specifications

Chemical Name or Material 2-Chloro-N-[4-[2,2,2-trifluoro-1-hydroxy-1-(trifluoromethyl)ethyl]phenyl]benzamide
CAS 303126-97-8
Quantity 10 mg
Target Nuclear Receptor Agonists
Molecular Formula C16H10ClF6NO2
Formula Weight 397.7
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.