Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SR 13800
GREENER_CHOICE

Catalog No. 543110
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
543110 10 mg
543150 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 543110 Supplier Tocris Bioscience™ Supplier No. 5431/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent MCT1 inhibitor

SR 13800 is a potent MCT1 inhibitor (IC50 = 0.5 nM); blocks proliferation of Raji lymphoma cells in vitro. Also inhibits lactate uptake in breast cancer cells in vitro.

Specifications

Chemical Name or Material 2,6-Dihydro-7-[(3-hydroxypropyl)thio]-2-methyl-4-(2-methylpropyl)-6-(1-naphthalenylmethyl)-1H-pyrrolo[3,4-d]pyridazin-1one
CAS 227321-12-2
Quantity 10 mg
Target Monocarboxylate Transporter Inhibitors
Molecular Formula C25H29N3O2S
Formula Weight 435.58
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.