Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SR 16832
GREENER_CHOICESDP

Catalog No. 63835 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
63-835 5 mg
63-832-5 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 63-835 Supplier Tocris Bioscience™ Supplier No. 6383/5
Add to Cart
Add to Cart
This item is not returnable. View return policy

PPAR gamma Receptor Antagonists

SR 16832 is a dual site PPARγ inhibitor. Acts at orthosteric and allosteric sites in the ligand binding domain. Inhibits binding of endogenous ligands and transcriptional activity of PPARγ, more effectively than the orthosteric covalent antagonists GW 9662 (Cat. No. 1508) and T 0070907 (Cat. No. 2301).

Specifications

CAS 2088135-12-8
Synonym 2-Chloro-N-(6-methoxy-4-quinolinyl)-5-nitrobenzamide
Quantity 5 mg
Molecular Formula C17H12ClN3O4
Formula Weight 357.75
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.