Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SR 2211
GREENER_CHOICE

Catalog No. p-4816764
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-691-0 10 mg
48-695-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 48-691-0 Supplier Tocris Bioscience™ Supplier No. 4869/10
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective RORγ inverse agonist

Selective inverse agonist of retinoic acid receptor-related orphan receptor γ (RORγ) (Ki = 105 nM; IC50 ∽320 nM). Exhibits weak activity at liver X receptor α (LXRα); does not affect the transcriptional activity of farnesoid X receptors (FXR), and has no impact on the transcriptional activity of RORα.

Chemical Identifiers

CAS 1359164-11-6
Molecular Formula C26H24F7N3O

Specifications

Quantity 10 mg
Formula Weight 527.48
Purity >98%
Molecular Weight (g/mol) 527.487
Chemical Name or Material SR 2211
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.