Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SR 3677 dihydrochloride

Catalog No. 366710R
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-671-0R 10 mg
36-675-0R 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 36-671-0R Supplier Tocris Bioscience™ Supplier No. 3667/10
This item is not returnable. View return policy

Potent, selective Rho-kinase (ROCK) inhibitor

SR 3677 dihydrochloride is a potent and selective Rho-kinase inhibitor (IC50 values are 3 and 56 nM for ROCK-II and ROCK-I respectively).

Specifications

Quantity 10 mg
Product Type SR 3677 dihydrochloride
Purity >98%
Molecular Weight (g/mol) 481.37
Inhibitors SR 3677 dihydrochloride

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.