Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SR 58611A hydrochloride
GREENER_CHOICESDP

Catalog No. 398610 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-861-0 10 mg
39-865-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 39-861-0 Supplier Tocris Bioscience™ Supplier No. 3986/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

β3 agonist; displays anxiolytic and antidepressant effects

Selective β3-adrenergic receptor agonist. Displays both anxiolytic and antidepressant effects in rodent models. Orally active and brain penetrant.

Chemical Identifiers

CAS 121524-09-2
Molecular Formula C22H27Cl2NO4

Specifications

Quantity 10 mg
Formula Weight 440.36
Purity >98%
Molecular Weight (g/mol) 440.361
Chemical Name or Material SR 58611A hydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.