Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SR 59230A hydrochloride
GREENER_CHOICESDP

Catalog No. 151110 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-111-0 10 mg
15-115-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 15-111-0 Supplier Tocris Bioscience™ Supplier No. 1511/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective β3 antagonist

SR 59230A hydrochloride is a potent and selective β3 adrenoceptor antagonist (IC50 values are 40, 408 and 648 nM for β3, β1 and β2 receptors respectively). Orally active in vivo.

Chemical Identifiers

CAS 1135278-41-9
Molecular Formula C21H28ClNO2

Specifications

Quantity 10 mg
Formula Weight 361.91
Purity >98%
Molecular Weight (g/mol) 361.91
Chemical Name or Material SR 59230A hydrochloride
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.