Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ SR 8278
GREENER_CHOICESDP

Catalog No. 446350 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-635-0 50 mg
44-631-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 44-635-0 Supplier R&D Systems™ Supplier No. 4463/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Rev-Erbα antagonist

Rev-Erbα antagonist; inhibits Rev-Erbα transcriptional repression (EC50 = 0.47 μM). Blocks activity of Rev-Erbα agonist GSK 4112 (Cat. No. 3663) in HEK293 cells. Increases expression of glucose-regulating genes, G6Pase and PEPCK in HepG2 cells.

Chemical Identifiers

CAS 1254944-66-5
Molecular Formula C18H19NO3S2

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 361.48
Purity >99%
Molecular Weight (g/mol) 361.474
Chemical Name or Material SR 8278
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.