Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SR 9009
GREENER_CHOICESDP

Catalog No. 585550 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
58-555-0 50 mg
58-551-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 58-555-0 Supplier Tocris Bioscience™ Supplier No. 5855/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rev-Erbα/β agonist

SR 9009 is a Rev-Erbα/β agonist (EC50 = 710 nM; Kd = 800 nM). Alters circadian locomotor activity and core clock hypothalamic gene expression in mice. Induces weight loss in diet-induced obese mice. Brain penetrant.

Specifications

Chemical Name or Material Ethyl 3-[[[(4-chlorophenyl)methyl][(5-nitro-2-thienyl)methyl]amino]methyl]-1-pyrrolidinecarboxylate
CAS 1379686-30-2
Quantity 50 mg
Target Nuclear Receptor Agonists
Molecular Formula C20H24ClN3O4S
Formula Weight 437.94
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.