Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ SR 9243
GREENER_CHOICESDP

Catalog No. 613850 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
61-385-0 50 mg
61-381-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 61-385-0 Supplier R&D Systems™ Supplier No. 6138/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective LXR inverse agonist

SR 9243 is a selective LXR inverse agonist. Exhibits no significant activity at other nuclear receptors at maximally effective concentration. Inhibits the Warburg effect and lipogenesis by down-regulation of LXR-mediated gene expression. Selectively reduces cell viability and induces apoptosis in cancer cell lines in vitro. Suppresses hepatic steatosis and inhibits growth of tumor xenografts in mice.

Specifications

CAS 1613028-81-1
Quantity 50 mg
Solubility Soluble to 20mM in DMSO
Product Line Tocris
Molecular Formula C31H32BrNO4S2
Formula Weight 626.62g/mol
Color White
Product Type Inhibitor

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.