Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ SR 95531 hydrobromide
GREENER_CHOICESDP

Catalog No. 126250 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-625-0 50 mg
12-621-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 12-625-0 Supplier R&D Systems™ Supplier No. 1262/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Competitive and selective GABAA antagonist

SR 95531 hydrobromide is a selective, competitive GABAA receptor antagonist and allosteric inhibitor (IC50 = 200 nM). SR 95531 displaces [3H]-GABA from rat brain membranes with a Ki of 150 nM. Unlike Bicuculline (Cat. No. 0130), SR 95531 selectively antagonizes GABA-induced Cl- currents with little action on Pentobarbitone -induced currents (Cat. No. 4579). SR 95531 can act as an agonist at high concentrations (>100 μM) and is a weak agonist at GABAA receptors with α1β2(Y157S)γ2L subunits. SR 95531 also acts as a competitive antagonist of recombinant glycine receptors and exhibits greater potency at glycine receptors containing β subunits.

Chemical Identifiers

CAS 104104-50-9
Molecular Formula C15H18BrN3O3

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 368.23
Purity >98%
Molecular Weight (g/mol) 368.231
Chemical Name or Material SR 95531 hydrobromide
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.