Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SR-AI/MSR Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB400159APC
Description
SR-AI/MSR Polyclonal antibody specifically detects SR-AI/MSR in Human, Primate samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
SR-AI/MSR | |
Polyclonal | |
Western Blot, Immunocytochemistry/ Immunofluorescence | |
CD204, CD204 antigen, Macrophage acetylated LDL receptor I and II, macrophage scavenger receptor 1, macrophage scavenger receptor type III, phSR1, phSR2, SCARA1macrophage scavenger receptor types I and II, Scavenger receptor class A member 1, scavenger receptor class A, member 1, SR-A | |
A synthetic peptide made to an internal portion of human Macrophage Scavenger Receptor I (between residues 300-350). [UniProt P21757] | |
0.1 mL | |
Adaptive Immunity, Cancer, Immunology, Innate Immunity, Lipid and Metabolism, Neuroscience, Prostate Cancer | |
4481 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunocytochemistry | |
APC | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Primate | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction