Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SR-AI/MSR Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP100092H
Description
SR-AI/MSR Polyclonal specifically detects SR-AI/MSR in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
SR-AI/MSR | |
Polyclonal | |
Western Blot, Immunohistochemistry-Frozen | |
P21757 | |
MSR1 | |
Synthetic peptide made to an internal portion of human Macrophage Scavenger Receptor I (within residues 400-450). [Swiss-Prot: P21757] | |
Affinity Purified | |
RUO | |
Primary | |
Reacts with human and mouse. 100% sequence identity with horse and dog, 93% identity with monkey and bovine, 92% identity with rat. | |
Store at 4C in the dark. |
Western Blot, Immunohistochemistry (Frozen) | |
HRP | |
PBS with No Preservative | |
CD204, CD204 antigen, Macrophage acetylated LDL receptor I and II, macrophage scavenger receptor 1, macrophage scavenger receptor type III, phSR1, phSR2, SCARA1macrophage scavenger receptor types I and II, Scavenger receptor class A member 1, scavenger receptor class A, member 1, SR-A | |
Rabbit | |
50 kDa | |
0.1 mL | |
Adaptive Immunity, Cancer, Immunology, Innate Immunity, Lipid and Metabolism, Neuroscience, Prostate Cancer | |
4481 | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction