Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SR-BI Antibody - BSA Free, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$269.00 - $562.50

Specifications

Antigen SR-BI
Concentration 1.0 mg/mL
Dilution Western Blot 1:1000 - 1:5000, Simple Western 1:100, Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 2.5 - 5 ug/mL, Immunocytochemistry/Immunofluorescence 1:50 - 1:1000, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 2.5 - 5 ug/mL, Immunohistochemistry-Frozen reported in scientific literature (PMID 26865459), Proximity Ligation Assay reported in scientific literature (10.1016/j.jbc.2021.100828), Knockout Validated reported in scientific literature (PMID 31462534), KnockDown Validated, Block/Neutralize reported in scientific literature (PMID 24859737)
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), In situ PLA, Immunoassay, KnockDown, Neutralization
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB400104SS
SDP
View Documents
Novus Biologicals
NB400104SS
0.025 mL
Each for $269.00
Only null left
Add to Cart
 
NB400104
SDP
View Documents
Novus Biologicals
NB400104
0.1 mL
Each for $562.50
Only null left
Add to Cart
 
Description

Description

SR-BI Polyclonal specifically detects SR-BI in Human, Mouse, Rat, Porcine, Chinese Hamster, Hamster, Mustelid, Primate, Rabbit, Golden Syrian Hamster samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Proximity Ligation Assay, Knockout Validated, Knockdown Validated, Block/Neutralize.
Specifications

Specifications

SR-BI
Western Blot 1:1000 - 1:5000, Simple Western 1:100, Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 2.5 - 5 ug/mL, Immunocytochemistry/Immunofluorescence 1:50 - 1:1000, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 2.5 - 5 ug/mL, Immunohistochemistry-Frozen reported in scientific literature (PMID 26865459), Proximity Ligation Assay reported in scientific literature (10.1016/j.jbc.2021.100828), Knockout Validated reported in scientific literature (PMID 31462534), KnockDown Validated, Block/Neutralize reported in scientific literature (PMID 24859737)
Polyclonal
Rabbit
Breast Cancer, Cancer, Cardiovascular Biology, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction, Virology Bacteria and Parasites
PBS with 0.1% Sodium Azide
CD36 and LIMPII analogous 1, CD36 antigen, CD36 antigen (collagen type I receptor, thrombospondin receptor)-like 1, CD36L1scavenger receptor class B type 1, CLA1 CD36 antigen-like 1, CLA-1 HDLQTL6, Collagen type I receptor, thrombospondin receptor-like 1, MGC138242, SCARB1, scavenger receptor class B type III, scavenger receptor class B, member 1, SR-B1, SRB1 scavenger receptor class B member 1, SRBI
SCARB1
IgG
Affinity Purified
Mouse, rat, mink, hamster, primate, rabbit and the long form of human SR-BI. Reactivity with CHO cells reported in scientific literature (PMID: 12356718).
1.0 mg/mL
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), In situ PLA, Immunoassay, KnockDown, Neutralization
Unconjugated
RUO
Human, Mouse, Rat, Chinese Hamster, Hamster, Mustelid, Primate, Rabbit, Golden Syrian Hamster
Q61009
949
A C-terminal peptide containing residues from mouse SR-BI (within residues 450-509). [UniProt# Q61009]
Primary
Store at 4C. Do not freeze.
82 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.