Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SR-BI Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB400104MFV610
Description
SR-BI Polyclonal antibody specifically detects SR-BI in Human, Mouse, Rat, Porcine, Chinese Hamster, Hamster, Mustelid, Primate, Rabbit, Golden Syrian Hamster samples. It is validated for Flow CytometrySpecifications
SR-BI | |
Polyclonal | |
Flow Cytometry | |
CD36 and LIMPII analogous 1, CD36 antigen, CD36 antigen (collagen type I receptor, thrombospondin receptor)-like 1, CD36L1scavenger receptor class B type 1, CLA1 CD36 antigen-like 1, CLA-1 HDLQTL6, Collagen type I receptor, thrombospondin receptor-like 1, MGC138242, SCARB1, scavenger receptor class B type III, scavenger receptor class B, member 1, SR-B1, SRB1 scavenger receptor class B member 1, SRBI | |
A C-terminal peptide containing residues from mouse SR-BI (within residues 450-509). [UniProt Q61009] | |
0.1 mL | |
Breast Cancer, Cancer, Cardiovascular Biology, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction, Virology Bacteria and Parasites | |
949 | |
Store at 4C in the dark. | |
IgG |
Flow Cytometry | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Pig, Chinese Hamster, Hamster, Mustelid | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction