Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SR-BI/SR-BII Polyclonal Antibody
GREENER_CHOICE

Catalog No. APSA6805
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
APSA6805 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. APSA6805 Supplier Invitrogen™ Supplier No. PA316805
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SR-BI/SR-BII Polyclonal Antibody for Western Blot, IHC (P), ICC/IF, Flow, IP

High density lipoproteins (HDLs) play a critical role in cholesterol metabolism and their plasma concentrations are inversely correlated with risk for atherosclerosis. SR-BI and SR-BII (previously known as SR-BI.2) are the alternatively spliced products of a single gene. SR-BII and SR-BI are identical except for the encoded c-terminal cytoplasmic domain. Both SR-BI and SR-BII bind HDL and mediates selective uptake of HDL cholesteryl ester, but with SR-BII having an approximately 4-fold lower efficiency than SR-BI. SR-BI and SR-BII are expressed primarily in liver and non-placental steroidgenic tissues. Although the role of these scavenger receptors is not completely clear, SR-BII mRNA results from the alternative splicing of SR-BI precursor transcripts with both isoforms mediating selective transfer of lipid between HDL and cells. Therefore, the relative expression and functional activities of these two isoforms create a potential means of regulating selective lipid transfer between HDL and cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SR-BI/SR-BII
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Whole serum with 0.02% sodium azide
Gene SCARB2
Gene Accession No. O35114, P27615, P97943, Q14108, Q61009, Q8WTV0
Gene Alias 85 kDa lysosomal membrane sialoglycoprotein; 85 kDa lysosomal sialoglycoprotein scavenger receptor class B, member 2; 9330185J12Rik; AI120173; AMRF; CD36; CD36 and LIMPII analogous 1; CD36 antigen (collagen type I receptor thrombospondin receptor)-like 1 (scavanger receptor class B type 1); CD36 antigen (collagen type I receptor, thrombospondin receptor)-like 1; CD36 antigen (collagen type I receptor, thrombospondin receptor)-like 2; CD36 antigen (collagen type I receptor, thrombospondin receptor)-like 2 (lysosomal integral membrane protein II); CD36 antigen-like 1; CD36 antigen-like 2; CD36L1; CD36L2; Cla1; CLA-1; Collagen type I receptor, thrombospondin receptor-like 1; D5Ertd460e; EPM4; HDL QTL 1; Hdlq1; HDLQTL6; Hlb398; HLGP85; LGP85; LIMP II; LIMP2; LIMP-2; LIMPII; lysosome membrane protein 2; Lysosome membrane protein II; MLGP85; mSR-BI; Scarb1; Scarb2; scavanger receptor class B type 1; scavenger receptor b1; scavenger receptor class B member 1; Scavenger receptor class B member 2; scavenger receptor class B type I; scavenger receptor class B type III; scavenger receptor class B, member 1; scavenger receptor class B, member 2; scavenger receptor class B1; SRB1; SR-B1; SR-B1/B2; SRBI; SR-BI; SR-BI/BII; SR-BII
Gene Symbols SCARB1, SCARB2
Host Species Rabbit
Immunogen A peptide from the extracellular domain (residues 230-380) of Scavenger Receptor-BI/BII that was expressed as two tandem copies in bacteria using the pET system.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117106, 12492, 20778, 25073, 949, 950
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.