Learn More
Stoelting™ SR-LAB™ Startle Response System
SDP
SR-LAB™ Startle Response System features a design that ensures accurate results for all paradigms and is ideal for startle reflex testing
$1611.00 - $31850.00
Specifications
For Use With (Application) Neuroscience
View More Specs
Products 7
Catalog Number Mfr. No. Model Price Quantity & Availability  
Catalog Number Mfr. No. Model Price Quantity & Availability  
10-001-054
SDP
View Documents
Stoelting™
59800
SR-LAB System for Desktop Computer
Each for $31,850.00
Only null left
Add to Cart
10-001-055
SDP
View Documents
Stoelting™
59802
SR-LAB System, Additional Unit
Each for $14,540.00
Only null left
Add to Cart
10-001-056
SDP
View Documents
Stoelting™
59825
SR-LAB S Animal Encl w/Ultrasensor
Each for $1,611.00
Only null left
Add to Cart
10-001-057
SDP
View Documents
Stoelting™
59826
SR-LAB S/M Animal Encl w/ULTsensor
Each for $1,651.00
Only null left
Add to Cart
10-001-058
SDP
View Documents
Stoelting™
59827
SR-LAB Medium Animal Enclosure
Each for $1,651.00
Only null left
Add to Cart
10-001-059
SDP
View Documents
Stoelting™
59828
SR-LAB Large Animal Enclosure
Each for $1,766.00
Only null left
Add to Cart
10-001-060
SDP
View Documents
Stoelting™
59829
SR-LAB X-Large Animal Enclosure
Each for $2,030.00
Only null left
Add to Cart
Description
Description
  • Easy to use with straightforward assembly and menu-driven software
  • Expands to include 16 test stations
  • Use in all startle paradigms including startle habituation, pre-pulse, and cross modal inhibition
  • Standardized test cabinets
  • Basic Startle Reflex hardware and Microsoft Windows based software give users complete control over startle paradigms. Virtually any combination of tones, noise bursts, lights, air puffs, background noise and foot shock options are controllable.
  • Supports up to 16 test stations.
  • Test cabinets allow full and unobtrusive observation of the test animals.
  • Data collected are completely secure and recorded in a non-editable form
SR-LAB Components basic system components include:
  • Test cabinet in composite ABS plastic
  • Animal enclosure in choice of standard sizes
  • Power Supply
  • Input and Output cables
  • Software and User Manual
  • Software utilities for data analysis
  • Animal Enclosures
  • Animal enclosures are designed to locate the subject without using restraint. The enclosure monitors movements with an ultra-stable, hermetically sealed motion sensor using new USB 16 bit resolution board (a range from zero to over 10,000 millivolts. A 16 bit board diveds 10,000 mV by 65536 discrete numbers. Thus, 10,000/65536 = 0.15 MV and now you get a new data point with just a 0.15 mV change) for measuring startle responses. All enclosures are adjustable in length and made of transparent acrylic for easy cleaning and convenient observation.
Software Features
The SR-Lab Software helps users plan, execute and manage data in startle reflex testing sessions. Users can define trial definitions and session definitions with a user-friendly interface. Test progress can be displayed with summary numeric data and response waveforms. After the session has ended, the complete waveform can be reviewed, and data may be refined via settable parameters including Baseline, Onset Window Start, End Analysis, and Onset criterion. The software stores all data in a database for easy exporting to Microsoft Excel and/or statistical software programs.
Specifications
Specifications
Neuroscience
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.