Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SRSF4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101148
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101148 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101148 Supplier Invitrogen™ Supplier No. PA5101148
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SFRS4.

Serine/arginine-rich protein 75 (SRp75) is encoded by the SRSF4 gene. SRp75 belongs to the arginine/serine-rich factor family and likely plays a role in alternative mRNA splicing.The family of SR factors all contain one or more RNA recognition motifs (RRM) and an arginine/ serine (RS)-rich domain. They are not only essential for constitutive splicing but also regulate splicing in a concentration-dependent manner by influencing the selection of alternative splice sites. The majority of SR proteins, including SC35 and SRp40, are confined to the nucleus, while SF2/ASF, SRp20, and 9G8 are continuously shuttled between the nucleus and the cytoplasm and contribute to mRNA transport. The activity of SR proteins in regulated splicing is antagonized by members of the hnRNP A/B family of proteins, which induce drastic shifts in the selection of splicing sites.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRSF4
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SRSF4
Gene Accession No. Q08170, Q8VE97
Gene Alias 5730499P16Rik; AW550192; MNCb-2616; Pre-mRNA-splicing factor SRP75; RGD1561347; serine and arginine rich splicing factor 4; serine/arginine-rich splicing factor 4; SFRS4; splicing factor, arginine/serine-rich 4; splicing factor, arginine/serine-rich 4 (SRp75); SR splicing factor 4; SRP001LB; SRP75; SRSF4
Gene Symbols SRSF4
Host Species Rabbit
Immunogen A synthesized peptide derived from human SRSF4(Accession Q08170), corresponding to amino acid residues R114-E164.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57317, 6429
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.