Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SRSF6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103322
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103322 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103322 Supplier Invitrogen™ Supplier No. PA5103322
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SFRS6.

SFRS6 is involved in mRNA splicing and may play a role in the determination of alternative splicing. It belongs to the splicing factor SR family and has been shown to bind with and modulate another member of the family, SFRS12. The protein encoded by this gene is involved in mRNA splicing and may play a role in the determination of alternative splicing. The encoded nuclear protein belongs to the splicing factor SR family and has been shown to bind with and modulate another member of the family, SFRS12.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRSF6
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Srsf6
Gene Accession No. G3V6S8, Q13247, Q3TWW8
Gene Alias 1210001E11Rik; AI314910; AW146126; B52; epididymis secretory protein Li 91; HEL-S-91; LOW QUALITY PROTEIN: serine/arginine-rich splicing factor 6; MGC5045; pre-mRNA-splicing factor SRP55; serine and arginine rich splicing factor 6; serine/arginine-rich splicing factor 6; serine/arginine-rich splicing factor 6-like protein; Sfrs6; splicing factor SRp55; splicing factor SRp55 homolog; splicing factor SRp55-1; splicing factor, arginine/serine-rich 6; splicing factor, arginine/serine-rich, 55 kDa; SR splicing factor 6; SRP55; SRP55-1; SRSF6
Gene Symbols Srsf6
Host Species Rabbit
Immunogen A synthesized peptide derived from human SRSF6(Accession Q13247), corresponding to amino acid residues S228-S278.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 362264, 6431, 67996
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.