Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SR Recombinant Rabbit Monoclonal Antibody (JE61-82)
GREENER_CHOICE

Catalog No. PIMA544854
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544854 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA544854 Supplier Invitrogen™ Supplier No. MA544854
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

SR Recombinant Monoclonal Antibody for Western Blot, IHC (P)

SR proteins are a highly conserved family of arginine/serine-rich, spliceosome-associated phosphoproteins essential for metazoan pre-mRNA splicing. SR proteins act early in splicing by promoting splice site recognition and spliceosome assembly. SR proteins also play a regulatory role because they can determine alternative splice site usage in vivo and in vitro. SR proteins appear to be recruited from nuclear speckles, in which they are concentrated to sites of transcription in order to spatially coordinate transcription and pre-mRNA splicing within the cell nucleus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SR
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone JE61-82
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene SRSF6
Gene Accession No. G3V6S8, O35326, P84103, P84104, Q08170, Q09167, Q13243, Q13247, Q3TWW8, Q8VE97
Gene Alias 1210001E11Rik; 5730499P16Rik; AI314910; AL024116; AW146126; AW550192; B52; Cl-4; Delayed-early protein HRS; epididymis secretory protein Li 91; HEL-S-91; HRS; hypothetical protein; insulin-induced growth response protein CL-4; LOW QUALITY PROTEIN: serine/arginine-rich splicing factor 4; MNCb-2616; pre-mRNA splicing factor SRp20; pre-mRNA-splicing factor SRP20; Pre-mRNA-splicing factor SRP40; pre-mRNA-splicing factor SRP55; Pre-mRNA-splicing factor SRP75; Protein X16; RGD1561347; serine and arginine rich splicing factor 3; serine and arginine rich splicing factor 4; serine and arginine rich splicing factor 5; serine and arginine rich splicing factor 6; serine/arginine-rich splicing factor 3; Serine/arginine-rich splicing factor 4; serine/arginine-rich splicing factor 5; Serine/arginine-rich splicing factor 6; SFRS3; Sfrs4; Sfrs5; Sfrs6; splicing factor; splicing factor arginine/serine-rich 5 (SRp40 HRS); splicing factor SRp55; splicing factor SRp55 homolog; splicing factor SRp55-1; splicing factor, arginine/serine-rich 3; splicing factor, arginine/serine-rich 3 (SRp20); splicing factor, arginine/serine-rich 4; splicing factor, arginine/serine-rich 4 (SRp75); splicing factor, arginine/serine-rich 5; splicing factor, arginine/serine-rich 5 (SRp40, HRS); splicing factor, arginine/serine-rich 6; splicing factor, arginine/serine-rich, 20-kD; splicing factor, arginine/serine-rich, 55 kDa; SR splicing factor 4; SR splicing factor 5; SR splicing factor 6; SRP001LB; Srp20; SRP40; SRP55; SRP55-1; SRP75; SRSF3; SRSF4; SRSF5; Srsf6; X16
Gene Symbols SRSF3, SRSF4, SRSF5, SRSF6
Host Species Rabbit
Immunogen Synthetic peptide within human SR protein repeat aa 150-199/344.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20383, 20384, 29667, 361814, 362264, 362612, 57317, 6428, 6429, 6430, 6431, 67996
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.