Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SR140 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566388
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566388 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566388 Supplier Invitrogen™ Supplier No. PA566388
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IEFVVREGPMF EAMIMNREIN NPMFRFLFEN QTPAHVYYRW KLYSILQGDS PTKWRTEDFR MFKNGSFWRP PPLNPYLHGM S Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.

U2SURP (U2 SnRNP Associated SURP Domain Containing) is a Protein Coding gene. Diseases associated with U2SURP include Retinitis Pigmentosa 67. Among its related pathways are Gene Expression and mRNA Splicing - Major Pathway. Gene Ontology (GO) annotations related to this gene include nucleic acid binding and RNA binding. [GeneCards].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SR140
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene U2SURP
Gene Accession No. O15042
Gene Alias 140 kDa Ser/Arg-rich domain protein; 2610101N10Rik; AU023006; fSAPa; functional spliceosome-associated protein a; KIAA0332; mp:zf637-1-000918; RGD1307882; Ser/Arg-rich domain protein, 140 kDa; si:dkey-181m9.9; SR140; U2 associated SR140 protein; U2 snRNP associated SURP domain containing; U2 snRNP-associated SURP domain containing; U2 snRNP-associated SURP motif-containing protein; U2-associated protein SR140; u2-associated protein SR140-like protein; U2-associated SR140 protein; U2SURP; wu:fa03e02; wu:fb38b01; wu:fc75d03
Gene Symbols U2SURP
Host Species Rabbit
Immunogen Recombinant Human SR140. Recombinant protein control fragment (Product #RP-107061).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23350
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.