Learn More
Invitrogen™ SR140 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110536
Description
Immunogen sequence: GAPLEDVDGI PIDATPIDDL DGVPIKSLDD DLDGVPLDAT EDSKKNEPIF KVAPSKWEAV DESELEAQAV TTSKWELFDQ HEESEEEENQ NQEEESEDEE DTQSSKSEEH HLYSNPIKEE MTESKFSKYS EMSEEKRAKL REIELKVMKF QDELESGKRP KKPGQSFQEQ VEHYRDKLLQ REKEKELERE RERDKKDKEK LESRSKDKKE KDECTPTRKE RKRRHSTSPS PSRSSSGRRV KSPSPKSERS ERSERSHKES SRSRSSHKDS PRDVSKKAKR SPSGSRTPKR SRRSRSRSPK KSGKKSRSQS RSPHRSHKKS KKNKH.
U2SURP (U2 SnRNP Associated SURP Domain Containing) is a Protein Coding gene. Diseases associated with U2SURP include Retinitis Pigmentosa 67. Among its related pathways are Gene Expression and mRNA Splicing - Major Pathway. Gene Ontology (GO) annotations related to this gene include nucleic acid binding and RNA binding. [GeneCards].
Specifications
| SR140 | |
| Polyclonal | |
| Unconjugated | |
| U2SURP | |
| 140 kDa Ser/Arg-rich domain protein; 2610101N10Rik; AU023006; fSAPa; functional spliceosome-associated protein a; KIAA0332; mp:zf637-1-000918; RGD1307882; Ser/Arg-rich domain protein, 140 kDa; si:dkey-181m9.9; SR140; U2 associated SR140 protein; U2 snRNP associated SURP domain containing; U2 snRNP-associated SURP domain containing; U2 snRNP-associated SURP motif-containing protein; U2-associated protein SR140; u2-associated protein SR140-like protein; U2-associated SR140 protein; U2SURP; wu:fa03e02; wu:fb38b01; wu:fc75d03 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 23350, 315903, 67958 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.89 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O15042, Q6NV83 | |
| U2SURP | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 705-1029 of human U2SURP (NP_001073884.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.