Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SRA1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | SRA1 |
---|---|
Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
SRA1 Polyclonal specifically detects SRA1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
SRA1 | |
Polyclonal | |
Rabbit | |
Human | |
MGC87674, pp7684, SRA, SRAP, steriod receptor RNA activator 1, steriod receptor RNA activator protein, steroid receptor coactivator, steroid receptor RNA activator 1, steroid receptor RNA activator 1 (complexes with NCOA1), Steroid receptor RNA activator protein, STRAA1 | |
SRA1 | |
IgG | |
Affinity Purified |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
10011 | |
This antibody was developed against a recombinant protein corresponding to amino acids: RMALLVQELSSHRWDAADDIHRSLMVDHVTEVSQWMVGVKRLIAEKRSLFSEEAANEEKSAATAEKNHTIPGFQQAS | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title