Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SRA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562145
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562145 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562145 Supplier Invitrogen™ Supplier No. PA562145
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RMALLVQELS SHRWDAADDI HRSLMVDHVT EVSQWMVGVK RLIAEKRSLF SEEAANEEKS AATAEKNHTI PGFQQAS Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 77%.

This gene is involved in transcriptional coactivation by steroid receptors. There is currently data suggesting this gene encodes both a non-coding RNA that functions as part of a ribonucleoprotein complex and a protein coding mRNA. Increased expression of both the transcript and the protein is associated with cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRA1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SRA1
Gene Accession No. Q9HD15
Gene Alias AA959952; LOW QUALITY PROTEIN: steroid receptor RNA activator 1; MGC87674; pp7684; RGD:621148}; Sra; sra1; sra1 {ECO:0000312; SRAP; steroid receptor coactivator; steroid receptor RNA activator 1; steroid receptor RNA activator 1 (complexes with NCOA1); steroid receptor RNA activator protein; Straa1; Strra1; Unknown (protein for MGC:128615)
Gene Symbols SRA1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SRA1. Recombinant protein control fragment (Product #RP-96989).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10011
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.