Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SRA1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA575729
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
This gene is involved in transcriptional coactivation by steroid receptors. There is currently data suggesting this gene encodes both a non-coding RNA that functions as part of a ribonucleoprotein complex and a protein coding mRNA. Increased expression of both the transcript and the protein is associated with cancer.
Specifications
SRA1 | |
Polyclonal | |
Unconjugated | |
SRA1 | |
AA959952; LOW QUALITY PROTEIN: steroid receptor RNA activator 1; MGC87674; pp7684; RGD:621148}; Sra; sra1; sra1 {ECO:0000312; SRAP; steroid receptor coactivator; steroid receptor RNA activator 1; steroid receptor RNA activator 1 (complexes with NCOA1); steroid receptor RNA activator protein; Straa1; Strra1; Unknown (protein for MGC:128615) | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10011, 24068, 252891 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6QGW5, Q80VJ2, Q9HD15 | |
SRA1 | |
Synthetic peptide corresponding to residues in Human SRA1. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction