Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Src Antibody (21H5), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | Src |
---|---|
Clone | 21H5 |
Dilution | Western Blot 1:500-1:5000, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence 1:20-1:200 |
Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry |
Classification | Monoclonal |
Description
Src Monoclonal antibody specifically detects Src in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, ImmunocytochemistrySpecifications
Src | |
Western Blot 1:500-1:5000, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence 1:20-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
c-src, EC 2.7.10, EC 2.7.10.2, pp60c-src, Rous sarcoma, tyrosine kinase pp60c-src, tyrosine-protein kinase SRC-1, v-src avian sarcoma (Schmidt-Ruppin A-2) viral oncogene homolog, v-src sarcoma (Schmidt-Ruppin A-2) viral oncogene homolog (avian) | |
A synthesized peptide derived from Human Src [UniProt P12931] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
21H5 | |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Cancer Stem Cells, Phospho Specific, Protein Kinase, Signal Transduction, Transcription Factors and Regulators, Tyrosine Kinases | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
6714 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title