Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Src Antibody Pack, Novus Biologicals™
SDP

Catalog No. NB234295 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
5 Vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB234295 5 Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB234295 Supplier Novus Biologicals Supplier No. NBP320214
Only null left
Add to Cart
Add to Cart

Antibody Pack

Src antibody specifically detects Src in Human samples. It is validated for Western Blot

Specifications

Antigen Src
Applications Western Blot
Conjugate Unconjugated
Gene Alias c-src, EC 2.7.10, EC 2.7.10.2, pp60c-src, Rous sarcoma, tyrosine kinase pp60c-src, tyrosine-protein kinase SRC-1, v-src avian sarcoma (Schmidt-Ruppin A-2) viral oncogene homolog, v-src sarcoma (Schmidt-Ruppin A-2) viral oncogene homolog (avian)
Immunogen See individual datasheets.
Quantity 5 Vials
Regulatory Status RUO
Gene ID (Entrez) 6714
Target Species Human
Content And Storage Storage is content dependent.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.