Learn More
Promega SRC Kinase Enzyme System, 1mg
Catalog No. PRV2922 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV2922 1 mg
1 options
Catalog No. PRV2922 Supplier Promega Supplier No. V2922
Add to Cart
Add to Cart
Request a Quote
Recombinant full-length human SRC was expressed in E. coli cells using an N-terminal GST tag. SRC was originally identified as a transforming protein of the Rous sarcoma virus that had enzymatic ability to phosphorylate tyrosine in protein substrates.
  • Easily Screen and Profile SRC Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C