Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Src Mouse anti-Avian, Human, Mouse, Rat, Mink, Clone: GD11, MilliporeSigma™

Catalog No. 05184I100UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05184I100UL 100 μL
05184I25UL 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 05184I100UL Supplier MilliporeSigma Supplier No. 05184I100UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Src Monoclonal antibody specifically detects Src in Avian, Human, Mouse, Rat, Mink samples. It is validated for Immunoprecipitation, IP-Kinase Assay, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Src
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone GD11
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Host Species Mouse
Immunogen A recombinant fragment expressed in E.coli corresponding to 88 amino acids from the N-terminal half of human Proto-oncogene tyrosine-protein kinase Src (c-Src).
Purification Method Protein G purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Avian, Human, Mouse, Rat, Mink
Content And Storage Stable for 1 year at +2°C to +8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.