Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SRC Rabbit anti-Human, Mouse, SureLight 488, Clone: C4, Abnova™

Catalog No. 89335930 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-335-930 10 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-335-930 Supplier Abnova Corporation Supplier No. MAB19096
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

SRC Monoclonal antibody specifically detects SRC in Human, Mouse samples. It is validated for Flow Cyt.

Specifications

Antigen SRC
Applications Flow Cytometry
Classification Monoclonal
Clone C4
Conjugate SureLight 488
Description v-src sarcoma (Schmidt-Ruppin A-2) viral oncogene homolog (avian)
Gene SRC
Gene Alias ASV, SRC1, c-SRC, p60-Src
Host Species Rabbit
Immunogen A synthetic phosphopeptide corresponding to residues surrounding Tyr416 of human phospho SRC
Quantity 10 Reactions
Primary or Secondary Primary
Gene ID (Entrez) 6714
Target Species Human, Mouse
Content And Storage Store at 4°C
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.