Learn More
Invitrogen™ Human SRC, His Tag Recombinant Protein

Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length, Recombinant, Cytoplasmic.

Specifications
Specifications
Accession Number | P12931 |
Concentration | See Label |
For Use With (Application) | Kinase Assay |
Formulation | 50mM tris with 1mM DTT, 0.05mM EDTA, 50% glycerol, 100mM NaCl, 0.05% NP-40 and no preservative; pH 7.5 |
Gene ID (Entrez) | 6714 |
Molecular Weight (g/mol) | 62.3 kDa |
Name | Human SRC, His Tag |
pH Range | 7.5 |
Purification Method | Purified |
Quantity | 10 μg |
Show More |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.