Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SRC1 Antibody, DyLight 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100312UV
Description
SRC1 Polyclonal antibody specifically detects SRC1 in Human, Mouse samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
SRC1 | |
Polyclonal | |
Western Blot, Immunohistochemistry | |
bHLHe42, BHLHE74, Class E basic helix-loop-helix protein 74, EC 2.3.1.48, Hin-2 protein, KAT13A, MGC129719, NCoA-1, nuclear receptor coactivator 1, PAX3/NCOA1 fusion protein, Protein Hin-2, Renal carcinoma antigen NY-REN-52, RIP160bHLHe74F-SRC-1, SRC-1, SRC1MGC129720, Steroid receptor coactivator 1, steroid receptor coactivator-1 | |
A 29 amino acid synthetic peptide, which represented a portion of human nuclear receptor coactivator 1 encoded within exon 12 (LocusLink ID 8648). | |
0.1 mL | |
Cancer, Chromatin Research, Transcription Factors and Regulators | |
8648 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry | |
DyLight 350 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction